missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Beta-1 adrenergic receptor Protein
A DNA sequence encoding the Homo sapiens (Human) Beta-1 adrenergic receptor, was expressed in the hosts and tags indicated.
287.00 EUR - 3175.00 EUR
Specifications
Accession Number | NP_000675.1 |
---|---|
Gene ID (Entrez) | 153 |
Name | Beta-1 adrenergic receptor Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16057301
|
enQuireBio™
QP8958-YE-100UG |
100 μg |
810.00 EUR
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16097301
|
enQuireBio™
QP8958-YE-500UG |
500 μg |
2325.00 EUR
500µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16067301
|
enQuireBio™
QP8958-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087301
|
enQuireBio™
QP8958-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16007311
|
enQuireBio™
QP8958-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16077301
|
enQuireBio™
QP8958-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000675.1 | |
Beta-1 adrenergic receptor Protein | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
153 | |
Research Use Only | |
ADRB1 | |
Recombinant Protein | |
Yeast | |
CRSPDFRKAF QRLLCCARRA ARRRHATHGD RPRASGCLAR PGPPPSPGAA SDDDDDDVVG ATPPARLLEP WAGCNGGAAA DSDSSLDEPC RPGFASESKV | |
Greater than 90% as determined by SDS-PAGE. |