missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Baculoviral IAP repeat-containing protein 1

A DNA sequence encoding the Homo sapiens (Human) Baculoviral IAP repeat-containing protein 8, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7945-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031761

  • 675.00 EUR / 100µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_203127.3
Baculoviral IAP repeat-containing protein 1
Research Use Only
BIRC8
Recombinant Protein
E. coli
MTGYEARLIT FGTWMYSVNK EQLARAGFYA IGQEDKVQCF HCGGGLANWK PKEDPWEQHA KWYPGCKYLL EEKGHEYINN IHLTRSLEGA LVQTTKKTPS LTKRISDTIF PNPMLQEAIR MGFDFKDVKK IMEERIQTSG SNYKTLGVLV ADLVSAQKDT TENELNQTSL QREISPEEPL RRLQEEKLCK ICMDRHIAVV FIPCGHLVTC KQCAEAVDRC PMCSMVIDFK QRVFMS
Greater than 80% as determined by SDS-PAGE.
112401
100 μg
Not determined.
Full Length of BC039318
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers