missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Baculoviral IAP repeat-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) Baculoviral IAP repeat-containing protein 8, was expressed in the hosts and tags indicated.
245.00 EUR - 2985.00 EUR
Specifications
Accession Number | NP_203127.3 |
---|---|
Gene ID (Entrez) | 112401 |
Name | Baculoviral IAP repeat-containing protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16031761
|
enQuireBio™
QP7945-EC-100UG |
100 μg |
675.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16051761
|
enQuireBio™
QP7945-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16041761
|
enQuireBio™
QP7945-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061761
|
enQuireBio™
QP7945-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16071761
|
enQuireBio™
QP7945-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081761
|
enQuireBio™
QP7945-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_203127.3 | |
Baculoviral IAP repeat-containing protein 1 | |
Not determined. | |
Full Length of BC039318 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
112401 | |
Research Use Only | |
BIRC8 | |
Recombinant Protein | |
E. coli | |
MTGYEARLIT FGTWMYSVNK EQLARAGFYA IGQEDKVQCF HCGGGLANWK PKEDPWEQHA KWYPGCKYLL EEKGHEYINN IHLTRSLEGA LVQTTKKTPS LTKRISDTIF PNPMLQEAIR MGFDFKDVKK IMEERIQTSG SNYKTLGVLV ADLVSAQKDT TENELNQTSL QREISPEEPL RRLQEEKLCK ICMDRHIAVV FIPCGHLVTC KQCAEAVDRC PMCSMVIDFK QRVFMS | |
Greater than 80% as determined by SDS-PAGE. |