missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ATP synthase subunit alpha, mitochondrial Protein

Product Code. 15987049
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Product Code. Quantity unitSize
15987049 100 μg 100µg
15997049 10 μg 10µg
15937059 50 μg 50µg
15917059 200 μg 200µg
15927059 500 μg 500µg
15907059 1 mg 1mg
6 options
This item is not returnable. View return policy

Product Code. 15987049

Brand: enQuireBio™ QP5693ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) ATP synthase subunit alpha, mitochondrial, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001001935.1
Gene ID (Entrez) 498
Name ATP synthase subunit alpha, mitochondrial Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol ATP5A1
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence QKTGTAEMSS ILEERILGAD TSVDLEETGR VLSIGDGIAR VHGLRNVQAE EMVEFSSGLK GMSLNLEPDN VGVVVFGNDK LIKEGDIVKR TGAIVDVPVG EELLGRVVDA LGNAIDGKGP IGSKTRRRVG LKAPGIIPRI SVREPMQTGI KAVDSLVPIG RGQRELIIGD RQTGKTSIAI DTIINQKRFN DGSDEKKKLY CIYVAIGQKR STVAQLVKRL TDADAMKYTI VVSATASDAA PLQYLAPYSG CSMGEYFRDN GKHALIIYDD LSKMS-QAVA YRQMSLLLRR PPGREAYPGD VFYLHSRLLE RAAKMNDAFG GGSLTALPVI ETQAGDVSAY IPTNVISITD GQIFLETELF YKGIRPAINV GLSVSRVGSA AQTRAMKQVA GTMKLELAQY REVAAFAQFG SDLDAATQQL LSRGVRLTEL LKQGQYSPMA IEEQVAVIYA GVRGYLDKLE PSKITKFENA FLSHVVSQHQ ALLGTIRADG KISEQSDAKL KEIVTNFLAG FEA
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.