missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Actin-like protein 7A Protein
A DNA sequence encoding the Homo sapiens (Human) Actin-like protein 7A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8283-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006678.1 | |
Actin-like protein 7A Protein | |
Research Use Only | |
ACTL7A | |
Recombinant Protein | |
E. coli | |
MWAPPAAIMG DGPTKKVGNQ APLQTQALQT ASLRDGPAKR AVWVRHTSSE PQEPTESKAA KERPKQEVTK AVVVDLGTGY CKCGFAGLPR PTHKISTTVG KPYMETAKTG DNRKETFVGQ ELNNTNVHLK LVNPLRHGII VDWDTVQDIW EYLFRQEMKI APEEHAVLVS DPPLSPHTNR EKYAEMLFEA FNTPAMHIAY QSRLSMYSYG RTSGLVVEVG HGVSYVVPIY EGYPLPSITG RLDYAGSDLT AYLLGLLNSA GNEFTQDQMG IVEDIKKKCC FVALDPIEEK KVPLSEHTIR YVLPDGKEIQ LCQERFLCSE MFFKPSLIKS MQLGLHTQTV SCLNKCDIAL KRDLMGNILL CGGSTMLSGF PNRLQKELSS MCPNDTPQVN VLPERDSAVW TGGSILASLQ GFQPLWVHRF EYEEHGPFFL YRRCF | |
Greater than 90% as determined by SDS-PAGE. |
10881 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |