missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Actin-like protein 7A Protein
A DNA sequence encoding the Homo sapiens (Human) Actin-like protein 7A, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_006678.1 |
---|---|
Gene ID (Entrez) | 10881 |
Name | Actin-like protein 7A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16013761
|
enQuireBio™
QP8283-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16033761
|
enQuireBio™
QP8283-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16023761
|
enQuireBio™
QP8283-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16043761
|
enQuireBio™
QP8283-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053761
|
enQuireBio™
QP8283-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16063761
|
enQuireBio™
QP8283-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_006678.1 | |
Actin-like protein 7A Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
10881 | |
Research Use Only | |
ACTL7A | |
Recombinant Protein | |
E. coli | |
MWAPPAAIMG DGPTKKVGNQ APLQTQALQT ASLRDGPAKR AVWVRHTSSE PQEPTESKAA KERPKQEVTK AVVVDLGTGY CKCGFAGLPR PTHKISTTVG KPYMETAKTG DNRKETFVGQ ELNNTNVHLK LVNPLRHGII VDWDTVQDIW EYLFRQEMKI APEEHAVLVS DPPLSPHTNR EKYAEMLFEA FNTPAMHIAY QSRLSMYSYG RTSGLVVEVG HGVSYVVPIY EGYPLPSITG RLDYAGSDLT AYLLGLLNSA GNEFTQDQMG IVEDIKKKCC FVALDPIEEK KVPLSEHTIR YVLPDGKEIQ LCQERFLCSE MFFKPSLIKS MQLGLHTQTV SCLNKCDIAL KRDLMGNILL CGGSTMLSGF PNRLQKELSS MCPNDTPQVN VLPERDSAVW TGGSILASLQ GFQPLWVHRF EYEEHGPFFL YRRCF | |
Greater than 90% as determined by SDS-PAGE. |