missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 60S ribosomal protein L31 Protein
A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L31, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8771-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000984.1 | |
60S ribosomal protein L31 Protein | |
Research Use Only | |
RPL31 | |
Recombinant Protein | |
E. coli | |
MAPAKKGGEK KKGRSAINEV VTREYTINIH KRIHGVGFKK RAPRALKEIR KFAMKEMGTP DVRIDTRLNK AVWAKGIRNV PYRIRVRLSR KRNEDEDSPN KLYTLVTYVP VTTFKNLQTV NVDEN | |
Greater than 90% as determined by SDS-PAGE. |
6160 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |