missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 60S ribosomal protein L31 Protein
A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L31, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_000984.1 |
---|---|
Gene ID (Entrez) | 6160 |
Name | 60S ribosomal protein L31 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16066391
|
enQuireBio™
QP8771-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16086391
|
enQuireBio™
QP8771-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16076391
|
enQuireBio™
QP8771-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16096391
|
enQuireBio™
QP8771-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16006401
|
enQuireBio™
QP8771-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16016401
|
enQuireBio™
QP8771-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000984.1 | |
60S ribosomal protein L31 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
6160 | |
Research Use Only | |
RPL31 | |
Recombinant Protein | |
E. coli | |
MAPAKKGGEK KKGRSAINEV VTREYTINIH KRIHGVGFKK RAPRALKEIR KFAMKEMGTP DVRIDTRLNK AVWAKGIRNV PYRIRVRLSR KRNEDEDSPN KLYTLVTYVP VTTFKNLQTV NVDEN | |
Greater than 90% as determined by SDS-PAGE. |