missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C virus genotype 1b (strain HC-J4) polyProtein
A DNA sequence encoding the Hepatitis C virus genotype 1a (isolate 1) (HCV) Hepatitis C virus genotype 1b (strain HC-J4) polyprotein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9739-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
Hepatitis C virus genotype 1b (strain HC-J4) polyProtein | |
Research Use Only | |
Partial (see sequence information for more details). | |
Hepatitis | |
His | |
Tris-based buffer, 50% glycerol |
10 μg | |
Not determined. | |
Recombinant Protein | |
Yeast | |
STNPKPQRKT KRNTNRRPQD VKFPGGGQIV GGVYLLPRRG PRLGVRATRK ASERSQPRGR RQPIPKARRP EGRAWAQPGY PWPLYGNEGL GWAGWLLSPR GSRPSWGPTD PRRRSRNLGK VIDTLTCGFA DLMGYIPLVG APLGGAARAL AHGVRVLEDG VNYATGNLPG CSFSIF | |
Greater than 90% as determined by SDS-PAGE. |