missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C virus genotype 1b (strain HC-J4) polyProtein
A DNA sequence encoding the Hepatitis C virus genotype 1a (isolate 1) (HCV) Hepatitis C virus genotype 1b (strain HC-J4) polyprotein, was expressed in the hosts and tags indicated.
352.00 EUR - 3175.00 EUR
Specifications
Name | Hepatitis C virus genotype 1b (strain HC-J4) polyProtein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Protein Length | Partial (see sequence information for more details). |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16008631
|
enQuireBio™
QP9739-YE-100UG |
100 μg |
965.00 EUR
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048631
|
enQuireBio™
QP9739-YE-500UG |
500 μg |
2325.00 EUR
500µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16018631
|
enQuireBio™
QP9739-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038631
|
enQuireBio™
QP9739-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058631
|
enQuireBio™
QP9739-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16028631
|
enQuireBio™
QP9739-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Hepatitis C virus genotype 1b (strain HC-J4) polyProtein | |
Not determined. | |
Recombinant Protein | |
Yeast | |
STNPKPQRKT KRNTNRRPQD VKFPGGGQIV GGVYLLPRRG PRLGVRATRK ASERSQPRGR RQPIPKARRP EGRAWAQPGY PWPLYGNEGL GWAGWLLSPR GSRPSWGPTD PRRRSRNLGK VIDTLTCGFA DLMGYIPLVG APLGGAARAL AHGVRVLEDG VNYATGNLPG CSFSIF | |
Greater than 90% as determined by SDS-PAGE. |
Research Use Only | |
Partial (see sequence information for more details). | |
Hepatitis | |
His | |
Tris-based buffer, 50% glycerol |