missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Uncharacterized protein YihF Protein
A DNA sequence encoding the Escherichia coli (strain K12) Uncharacterized protein YihF, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7139-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_418298.4 | |
Uncharacterized protein YihF Protein | |
Research Use Only | |
yihF | |
Recombinant Protein | |
E. coli | |
GTQIQPGVEK FIKDFNDAKK KGEHAYDMTL SYQNFDKGFF NSRFQMQMTF DNGAPDLNIK PGQKVVFDVD VEHGPLPITM LMHGNVIPAL AAAKVNLVNN ELTQPLFIAA KNKSPVEATL RFAFGGSFST TLDVAPAEYG KFSFGEGQFT FNGDGSSLSN LDIEGKVEDI VLQLSPMNKV TAKSFTIDSL ARLEEKKFPV GESESKFNQI NIINHGEDVA QIDAFVAKTR LDRVKDKDYI NVNLTYELDK LTKGNQQLGS GEWSLIAESI DPSAVRQFII QYNIAMQKQL AAHPELANDE VALQEVNAAL FKEYLPLLQK SEPTIKQPVR WKNALGELNA NLDISIADPA KSSSSTNKDI KSLNFDVKLP LNVVTETAKQ LNLSEGMDAE KAQKQADKQI SGMMTLGQMF QLITIDNNTA SLQLRYTPGK VVFNGQEMSE EEFMSRAGRF VH | |
Greater than 90% as determined by SDS-PAGE. |
948352 | |
1 mg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |