missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Uncharacterized protein YihF Protein
A DNA sequence encoding the Escherichia coli (strain K12) Uncharacterized protein YihF, was expressed in the hosts and tags indicated.
437.00 EUR - 2985.00 EUR
Specifications
Accession Number | NP_418298.4 |
---|---|
Gene ID (Entrez) | 948352 |
Name | Uncharacterized protein YihF Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16067480
|
enQuireBio™
QP7139-EC-100UG |
100 μg |
865.00 EUR
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16077480
|
enQuireBio™
QP7139-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16087480
|
enQuireBio™
QP7139-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16097480
|
enQuireBio™
QP7139-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16007490
|
enQuireBio™
QP7139-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_418298.4 | |
Uncharacterized protein YihF Protein | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
948352 | |
Research Use Only | |
yihF | |
Recombinant Protein | |
E. coli | |
GTQIQPGVEK FIKDFNDAKK KGEHAYDMTL SYQNFDKGFF NSRFQMQMTF DNGAPDLNIK PGQKVVFDVD VEHGPLPITM LMHGNVIPAL AAAKVNLVNN ELTQPLFIAA KNKSPVEATL RFAFGGSFST TLDVAPAEYG KFSFGEGQFT FNGDGSSLSN LDIEGKVEDI VLQLSPMNKV TAKSFTIDSL ARLEEKKFPV GESESKFNQI NIINHGEDVA QIDAFVAKTR LDRVKDKDYI NVNLTYELDK LTKGNQQLGS GEWSLIAESI DPSAVRQFII QYNIAMQKQL AAHPELANDE VALQEVNAAL FKEYLPLLQK SEPTIKQPVR WKNALGELNA NLDISIADPA KSSSSTNKDI KSLNFDVKLP LNVVTETAKQ LNLSEGMDAE KAQKQADKQI SGMMTLGQMF QLITIDNNTA SLQLRYTPGK VVFNGQEMSE EEFMSRAGRF VH | |
Greater than 90% as determined by SDS-PAGE. |