missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Methyl-accepting chemotaxis protein 1
A DNA sequence encoding the Escherichia coli (strain K12) Methyl-accepting chemotaxis protein I, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9632-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_418775.1 | |
Methyl-accepting chemotaxis protein 1 | |
Research Use Only | |
tsr | |
Recombinant Protein | |
Yeast | |
WFGIKASLVA PMNRLIDSIR HIAGGDLVKP IEVDGSNEMG QLAESLRHMQ GELMRTVGDV RNGANAIYSG ASEIATGNND LSSRTEQQAA SLEETAASME QLTATVKQNA ENARQASHLA LSASETAQRG GKVVDNVVQT MRDISTSSQK IADIISVIDG IAFQTNILAL NAAVEAARAG EQGRGFAVVA GEVRNLAQRS AQAAREIKSL IEDSVGKVDV GSTLVESAGE TMAEIVSAVT RVTDIMGEIA SASDEQSRGI DQVGLAVAEM DRVTQQNAAL VEESAAAAAA LEEQASRLTE AVAVFRIQQQ QRETSAVVKT VTPAAPRKMA VADSEENWET F | |
Greater than 90% as determined by SDS-PAGE. |
948884 | |
50 μg | |
Not determined. | |
Cytoplasmic Domain | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |