missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Methyl-accepting chemotaxis protein 1
A DNA sequence encoding the Escherichia coli (strain K12) Methyl-accepting chemotaxis protein I, was expressed in the hosts and tags indicated.
352.00 EUR - 3175.00 EUR
Specifications
Accession Number | NP_418775.1 |
---|---|
Gene ID (Entrez) | 948884 |
Name | Methyl-accepting chemotaxis protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16088531
|
enQuireBio™
QP9632-YE-100UG |
100 μg |
965.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16028541
|
enQuireBio™
QP9632-YE-500UG |
500 μg |
2325.00 EUR
500µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16098531
|
enQuireBio™
QP9632-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018541
|
enQuireBio™
QP9632-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038541
|
enQuireBio™
QP9632-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16008541
|
enQuireBio™
QP9632-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_418775.1 | |
Methyl-accepting chemotaxis protein 1 | |
Not determined. | |
Cytoplasmic Domain | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
948884 | |
Research Use Only | |
tsr | |
Recombinant Protein | |
Yeast | |
WFGIKASLVA PMNRLIDSIR HIAGGDLVKP IEVDGSNEMG QLAESLRHMQ GELMRTVGDV RNGANAIYSG ASEIATGNND LSSRTEQQAA SLEETAASME QLTATVKQNA ENARQASHLA LSASETAQRG GKVVDNVVQT MRDISTSSQK IADIISVIDG IAFQTNILAL NAAVEAARAG EQGRGFAVVA GEVRNLAQRS AQAAREIKSL IEDSVGKVDV GSTLVESAGE TMAEIVSAVT RVTDIMGEIA SASDEQSRGI DQVGLAVAEM DRVTQQNAAL VEESAAAAAA LEEQASRLTE AVAVFRIQQQ QRETSAVVKT VTPAAPRKMA VADSEENWET F | |
Greater than 90% as determined by SDS-PAGE. |