missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Ethanolamine ammonia-lyase light chain Protein
A DNA sequence encoding the Escherichia coli (strain K12) Ethanolamine ammonia-lyase light chain, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8927-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_416935.1 | |
Ethanolamine ammonia-lyase light chain Protein | |
Research Use Only | |
eutC | |
Recombinant Protein | |
E. coli | |
MDQKQIEEIV RSVMASMGQA APAPSEAKCA TTNCAAPVTS ESCALDLGSA EAKAWIGVEN PHRADVLTEL RRSTVARVCT GRAGPRPRTQ ALLRFLADHS RSKDTVLKEV PEEWVKAQGL LEVRSEISDK NLYLTRPDMG RRLCAEAVEA LKAQCVANPD VQVVISDGLS TDAITVNYEE ILPPLMAGLK QAGLKVGTPF FVRYGRVKIE DQIGEILGAK VVILLVGERP GLGQSESLSC YAVYSPRMAT TVEADRTCIS NIHQGGTPPV EAAAVIVDLA KRMLEQKASG INMTR | |
Greater than 90% as determined by SDS-PAGE. |
946925 | |
1 mg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |