missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Ethanolamine ammonia-lyase light chain Protein
A DNA sequence encoding the Escherichia coli (strain K12) Ethanolamine ammonia-lyase light chain, was expressed in the hosts and tags indicated.
309.00 EUR - 2645.00 EUR
Specifications
Accession Number | NP_416935.1 |
---|---|
Gene ID (Entrez) | 946925 |
Name | Ethanolamine ammonia-lyase light chain Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16087191
|
enQuireBio™
QP8927-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16027201
|
enQuireBio™
QP8927-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16077191
|
enQuireBio™
QP8927-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16007201
|
enQuireBio™
QP8927-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16017201
|
enQuireBio™
QP8927-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16097191
|
enQuireBio™
QP8927-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_416935.1 | |
Ethanolamine ammonia-lyase light chain Protein | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
946925 | |
Research Use Only | |
eutC | |
Recombinant Protein | |
E. coli | |
MDQKQIEEIV RSVMASMGQA APAPSEAKCA TTNCAAPVTS ESCALDLGSA EAKAWIGVEN PHRADVLTEL RRSTVARVCT GRAGPRPRTQ ALLRFLADHS RSKDTVLKEV PEEWVKAQGL LEVRSEISDK NLYLTRPDMG RRLCAEAVEA LKAQCVANPD VQVVISDGLS TDAITVNYEE ILPPLMAGLK QAGLKVGTPF FVRYGRVKIE DQIGEILGAK VVILLVGERP GLGQSESLSC YAVYSPRMAT TVEADRTCIS NIHQGGTPPV EAAAVIVDLA KRMLEQKASG INMTR | |
Greater than 90% as determined by SDS-PAGE. |