missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli 50S ribosomal protein L27 Protein
A DNA sequence encoding the Escherichia coli (strain K12) 50S ribosomal protein L27, was expressed in the hosts and tags indicated.
309.00 EUR - 2645.00 EUR
Specifications
Accession Number | NP_417652.1 |
---|---|
Gene ID (Entrez) | 945190 |
Name | 50S ribosomal protein L27 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16005701
|
enQuireBio™
QP8638-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16045701
|
enQuireBio™
QP8638-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16095691
|
enQuireBio™
QP8638-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16025701
|
enQuireBio™
QP8638-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16035701
|
enQuireBio™
QP8638-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16015701
|
enQuireBio™
QP8638-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_417652.1 | |
50S ribosomal protein L27 Protein | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
945190 | |
Research Use Only | |
rpmA | |
Recombinant Protein | |
E. coli | |
AHKKAGGSTR NGRDSEAKRL GVKRFGGESV LAGSIIVRQR GTKFHAGANV GCGRDHTLFA KADGKVKFEV KGPKNRKFIS IEAE | |
Greater than 90% as determined by SDS-PAGE. |