missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli 50S ribosomal protein L27 Protein
A DNA sequence encoding the Escherichia coli (strain K12) 50S ribosomal protein L27, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8638-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_417652.1 | |
50S ribosomal protein L27 Protein | |
Research Use Only | |
rpmA | |
Recombinant Protein | |
E. coli | |
AHKKAGGSTR NGRDSEAKRL GVKRFGGESV LAGSIIVRQR GTKFHAGANV GCGRDHTLFA KADGKVKFEV KGPKNRKFIS IEAE | |
Greater than 90% as determined by SDS-PAGE. |
945190 | |
100 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |