missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog MMP13 Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) MMP13 Protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6933-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
MMP13 Protein | |
Research Use Only | |
MMP13 | |
Recombinant Protein | |
E. coli | |
LPLPSDGDDD LSEEDFQLAE RYLKSYYYPL NPAGILKKSA AGSVADRLRE MQSFFGLEVT GKLDDNTLDI MKKPRCGVPD VGEYNVFPRT LKWSKTNLTY RIVNYTPDLT HSEVEKAFKK AFKVWSDVTP LNFTRLHDGT ADIMISFGTK EHGDFYPFDG PSGLLAHAFP PGPNYGGDAH FDDDETWTSS SKGYNLFLVA AHEFGHSLGL DHSKDPGALM FPIYTYTGKS HFMLPDDDVQ GIQSLYGPGD EDPNPRHPKT PDKCDPSLSL DAITSLRGET MIFKDRFFWR LHPQQVDAEL FLTKSFWPEL PNRIDAAYEH PSRDLIFIFR GRKYWALNGY DILEGYPQKI SELGFPKEVK KISAAVHFED TGKTLFFSGN QVWSYDDTNQ IMDKDYPRLI EEDFPGIGDK VDAVYEKNGY IYFFNGPIQF EYNIWSKRIV RVMPANSLLW C | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Full Length | |
Dog | |
His-SUMO | |
Tris-based buffer, 50% glycerol |