missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog MMP13 Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) MMP13 Protein, was expressed in the hosts and tags indicated.
309.00 EUR - 2985.00 EUR
Specifications
Name | MMP13 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | MMP13 |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16056750
|
enQuireBio™
QP6933-EC-100UG |
100 μg |
865.00 EUR
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16076750
|
enQuireBio™
QP6933-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16066750
|
enQuireBio™
QP6933-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16086750
|
enQuireBio™
QP6933-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16096750
|
enQuireBio™
QP6933-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16006760
|
enQuireBio™
QP6933-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
MMP13 Protein | |
Not determined. | |
Full Length | |
Dog | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
MMP13 | |
Recombinant Protein | |
E. coli | |
LPLPSDGDDD LSEEDFQLAE RYLKSYYYPL NPAGILKKSA AGSVADRLRE MQSFFGLEVT GKLDDNTLDI MKKPRCGVPD VGEYNVFPRT LKWSKTNLTY RIVNYTPDLT HSEVEKAFKK AFKVWSDVTP LNFTRLHDGT ADIMISFGTK EHGDFYPFDG PSGLLAHAFP PGPNYGGDAH FDDDETWTSS SKGYNLFLVA AHEFGHSLGL DHSKDPGALM FPIYTYTGKS HFMLPDDDVQ GIQSLYGPGD EDPNPRHPKT PDKCDPSLSL DAITSLRGET MIFKDRFFWR LHPQQVDAEL FLTKSFWPEL PNRIDAAYEH PSRDLIFIFR GRKYWALNGY DILEGYPQKI SELGFPKEVK KISAAVHFED TGKTLFFSGN QVWSYDDTNQ IMDKDYPRLI EEDFPGIGDK VDAVYEKNGY IYFFNGPIQF EYNIWSKRIV RVMPANSLLW C | |
Greater than 90% as determined by SDS-PAGE. |