Learn More
enQuireBio™ Recombinant Dog IL4 / Interleukin-4 Protein
Product Code. 16011700
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Product Code. 16011700
Brand: enQuireBio™ QP6222ec100ug
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) IL4 / Interleukin-4, was expressed in the hosts and tags indicated.
Specifications
Accession Number NP_001003159.1
Gene ID (Entrez) 403785
Name IL4 / Interleukin-4 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol IL4
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Dog
Species E. coli
Protein Tag His
Sequence HNFNITIKEI IKMLNILTAR NDSCMELTVK DVFTAPKNTS DKEIFCRAAT VLRQIYTHNC SNRYLRGLYR NLSSMANKTC SMNEIKKSTL KDFLERLKVI MQKKYYRH
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.