missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chicken DCX Protein
A DNA sequence encoding the Gallus gallus (Chicken) DCX, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8827-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001292603.1 | |
DCX Protein | |
Research Use Only | |
DCX | |
Recombinant Protein | |
E. coli | |
MELDFGHFDE RDKASRNMRG TRMNGLPSPT HSAHCSFYRT RTLQALSNEK KAKKVRFYRN GDRYFKGIVY AVSSDRFRSF DALLADLTRS LSDNINLPQG VRYIYTIDGS RKIGSMDELE EGESYVCSSD NFFKKVEYTK NVNPNWSVNV KTSANQKAPQ SLASSNSAQA KENKDFVRPK LVTIIRSGVK PRKAVRVLLN KKTAHSFEQV LTDITEAIKL ETGVVKKLYT LDGKQVTCLH DFFGDDDVFI ACGPEKFRYA QDDFSLDESE CRVMKGSPAA ATGSKSSPTP QKSSAKSPAP MRRSKSPADS ANGTSSSQLS TPKSKQSPIS TPTSPGSLRK HKVDLYLPLS LDDSDSLGDS M | |
Greater than 90% as determined by SDS-PAGE. |
374242 | |
1 mg | |
Not determined. | |
Full Length | |
Chicken | |
His | |
Tris-based buffer, 50% glycerol |