missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chicken DCX Protein
A DNA sequence encoding the Gallus gallus (Chicken) DCX, was expressed in the hosts and tags indicated.
309.00 EUR - 2985.00 EUR
Specifications
Accession Number | NP_001292603.1 |
---|---|
Gene ID (Entrez) | 374242 |
Name | DCX Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16096681
|
enQuireBio™
QP8827-EC-100UG |
100 μg |
865.00 EUR
100µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16016691
|
enQuireBio™
QP8827-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16006691
|
enQuireBio™
QP8827-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16026691
|
enQuireBio™
QP8827-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16036691
|
enQuireBio™
QP8827-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16046691
|
enQuireBio™
QP8827-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001292603.1 | |
DCX Protein | |
Not determined. | |
Full Length | |
Chicken | |
His | |
Tris-based buffer, 50% glycerol |
374242 | |
Research Use Only | |
DCX | |
Recombinant Protein | |
E. coli | |
MELDFGHFDE RDKASRNMRG TRMNGLPSPT HSAHCSFYRT RTLQALSNEK KAKKVRFYRN GDRYFKGIVY AVSSDRFRSF DALLADLTRS LSDNINLPQG VRYIYTIDGS RKIGSMDELE EGESYVCSSD NFFKKVEYTK NVNPNWSVNV KTSANQKAPQ SLASSNSAQA KENKDFVRPK LVTIIRSGVK PRKAVRVLLN KKTAHSFEQV LTDITEAIKL ETGVVKKLYT LDGKQVTCLH DFFGDDDVFI ACGPEKFRYA QDDFSLDESE CRVMKGSPAA ATGSKSSPTP QKSSAKSPAP MRRSKSPADS ANGTSSSQLS TPKSKQSPIS TPTSPGSLRK HKVDLYLPLS LDDSDSLGDS M | |
Greater than 90% as determined by SDS-PAGE. |