missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Decorin / DCN / SLRR1B Protein

A DNA sequence encoding the Bos taurus (Bovine) Decorin / DCN / SLRR1B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9101-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16007661

  • 2325.00 EUR / 500µg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_776331.2
Decorin / DCN / SLRR1B Protein
Research Use Only
DCN
Recombinant Protein
Yeast
DEASGIGPEE HFPEVPEIEP MGPVCPFRCQ CHLRVVQCSD LGLEKVPKDL PPDTALLDLQ NNKITEIKDG DFKNLKNLHT LILINNKISK ISPGAFAPLV KLERLYLSKN QLKELPEKMP KTLQELRVHE NEITKVRKSV FNGLNQMIVV ELGTNPLKSS GIENGAFQGM KKLSYIRIAD TNITTIPQGL PPSLTELHLD GNKITKVDAA SLKGLNNLAK LGLSFNSISA VDNGSLANTP HLRELHLNNN KLVKVPGGLA DHKYIQVVYL HNNNISAIGS NDFCPPGYNT KKASYSGVSL FSNPVQYWEI QPSTFRCVYV RAAVQLGNYK
Greater than 90% as determined by SDS-PAGE.
280760
500 μg
Not determined.
Full Length
Bovine
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers