missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Decorin / DCN / SLRR1B Protein
A DNA sequence encoding the Bos taurus (Bovine) Decorin / DCN / SLRR1B, was expressed in the hosts and tags indicated.
352.00 EUR - 3175.00 EUR
Specifications
Accession Number | NP_776331.2 |
---|---|
Gene ID (Entrez) | 280760 |
Name | Decorin / DCN / SLRR1B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16067651
|
enQuireBio™
QP9101-YE-100UG |
100 μg |
965.00 EUR
100µg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16007661
|
enQuireBio™
QP9101-YE-500UG |
500 μg |
2325.00 EUR
500µg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16077651
|
enQuireBio™
QP9101-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16097651
|
enQuireBio™
QP9101-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16017661
|
enQuireBio™
QP9101-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087651
|
enQuireBio™
QP9101-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_776331.2 | |
Decorin / DCN / SLRR1B Protein | |
Not determined. | |
Full Length | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |
280760 | |
Research Use Only | |
DCN | |
Recombinant Protein | |
Yeast | |
DEASGIGPEE HFPEVPEIEP MGPVCPFRCQ CHLRVVQCSD LGLEKVPKDL PPDTALLDLQ NNKITEIKDG DFKNLKNLHT LILINNKISK ISPGAFAPLV KLERLYLSKN QLKELPEKMP KTLQELRVHE NEITKVRKSV FNGLNQMIVV ELGTNPLKSS GIENGAFQGM KKLSYIRIAD TNITTIPQGL PPSLTELHLD GNKITKVDAA SLKGLNNLAK LGLSFNSISA VDNGSLANTP HLRELHLNNN KLVKVPGGLA DHKYIQVVYL HNNNISAIGS NDFCPPGYNT KKASYSGVSL FSNPVQYWEI QPSTFRCVYV RAAVQLGNYK | |
Greater than 90% as determined by SDS-PAGE. |