Learn More
enQuireBio™ Recombinant Bovine Acetylcholinesterase / ACHE Protein
Product Code. 15916399
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Product Code. 15916399
Brand: enQuireBio™ QP5606ec100ug
A DNA sequence encoding the Bos taurus (Bovine) Acetylcholinesterase / ACHE, was expressed in the hosts and tags indicated.
Specifications
Accession Number NP_001069688.1
Gene ID (Entrez) 540446
Name Acetylcholinesterase / ACHE Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol ACHE
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Bovine
Species E. coli
Protein Tag GST
Sequence EGPEDPELLV MVRGGRLRGL RLMAPRGPVS AFLGIPFAEP PVGPRRFLPP EPKRPWPGVL NATAFQSVCY QYVDTLYPGF EGTEMWNPNR ELSEDCLYLN VWTPYPRPSS PTPVLVWIYG GGFYSGASSL DVYDGRFLTQ AEGTVLVSMN YRVGAFGFLA LPGSREAPGN VGLLDQRLAL QWVQENVAAF GGDPTSVTLF GESAGAASVG MHLLSPPSRG LFHRAVLQSG APNGPWATVG VGEARRRATL LARLVGCPPG GAGGNDTELV ACLRARPAQD LVDHEWRVLP QESVFRFSFV PVVDGDFLSD TPEALINAGD FHGLQVLVGV VKDEGSYFLV YGAPGFSKDN ESLISRAQFL AGVRVGVPQA SDLAAEAVVL HYTDWLHPED PARLREALSD VVGDHNVVCP VAQLAGRLAA QGARVYAYIF EHRASTLSWP LWMGVPHGYE IEFIFGLPLE PSLNYTIEER TFAQRLMRYW ANFARTGDPN DPRDPKAPQW PPYTAGAQQY VSLNLRPLEV RRGLRAQACA FWNRFLPKLL SATDTLDEAE RQWKAEFHRW SSYMVHWKNQ FDHYSKQDRC SDL
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.