Learn More
Heidolph™ Coated Receiving Flask
Shop All Heidolph Instruments Products
Click to view available options
Volume (Metric):
100 mL
250 mL
500 mL
1000 mL
3000 mL
Unit Size:
Each
Description
- With S 35/20
- With transparent plastic coating
Specifications
Specifications
| Color | Clear |
| Material | Glass |
| Type | Receiving Flask |
| For Use With (Equipment) | Rotary evaporators |
| Material (Coating) | Transparent plastic |
| Standard Taper Joint Number Center | 35/20 |
| Volume (Metric) | 1000 mL |