missing translation for 'onlineSavingsMsg'
Learn More

Reboxetine mesylate, MedChemExpress

Product Code. 16442237
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16442237

Brand: Medchem Express HY14560C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Reboxetine mesylate (FCE20124 mesylate) is a potent, selective, and specific noradrenaline reuptake inhibitor (NARI) for the research of depression. Reboxetine mesylate inhibits the uptake of norepinephrine, with a Ki of 8 nM.

Reboxetine mesylate (FCE20124 mesylate) is a potent, selective, and specific noradrenaline reuptake inhibitor (NARI) for the research of depression. Reboxetine mesylate inhibits the uptake of norepinephrine, with a Ki of 8 nM.

Specifications

Chemical Name or Material Reboxetine mesylate
CAS 98769-84-7
Color White
Purity Grade Notes Research
Molecular Formula C20H27NO6S
Quantity 50 mg
Synonym FCE20124 mesylate PNU155950E mesylate
Solubility Information DMSO : 125 mg/mL (305.25 mM; Need ultrasonic) ∣H2O : 50 mg/mL (122.10 mM; Need ultrasonic)
SMILES CS(=O)(O)=O.CCOC1=C(C=CC=C1)O[C@]([C@@]2([H])CNCCO2)([H])C3=CC=CC=C3.[relative stereochemistry]
Molecular Weight (g/mol) 409.5
Formula Weight 409.5
Percent Purity 99.11%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.