Learn More
MedChemExpress Real Thiol
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Content And Storage | Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month |
| Quantity | 1 mg |
| Shipping Condition | Room temperature |
| CAS | 2280796-90-7 |
| Molecular Weight (g/mol) | 411.36 g/mol |
| Solubility | DMSO : ≥ 101 mg/mL (245.53 mM) |