Learn More
Cytiva Ready-To-Go™ GenomiPhi HY DNA Amplification Kit
Alle ansehen Cytiva Produkte
missing translation for 'orderingAttributeHoverText'
No. of Reactions:
24
96
missing translation for 'unitSize'
Each
Beschreibung
illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit is part of the Phi29 DNA polymerase family of products from Cytiva.
- Representative amplification of the whole genome.
- Preformulated, predispensed, single-dose, lyophilized cakes for rapid reaction set-up and greater reproducibility.
- Quick and simple automation-friendly protocol; no thermal cycler required.
- Shipping and storage at room temperature provides easy access and reduced carbon footprint.
- Yields high-quality DNA (e.g. for high-throughput genotyping and DNA archival).
Spezifikation
Spezifikation
| Content And Storage | Store the kit at ambient temperature.Once opened, completely reseal the pouch containingthe Ready-To-Go GenomiPhi V3 DNA amplification kit cakes. To ensure maximum shelf life and optimal performance, storage in a desiccator is recommended.Store recons |
| Description | illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 60 μg yield from nanogram amounts of DNA sample. |
| For Use With (Application) | Genomic DNA Amplification |
| Includes | 1 x 2.6mL 2X Denaturation Buffer, 3 x 96-well strips Ready-To-Go™ GenomiPhi HY cakes, 1 x vial control DNA, 12 strips of 8 domed caps |
| No. of Reactions | 96 |
| Product Type | illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit 96 Reactions |
| Size | > 10 kb |
| Test Time | 4 h |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur