Learn More
Applied Biosystems™ Reaction Vessel, 40 mL, RTF version
Shop All Applied Biosystems ProductsDescription
This new vessel uses tabbed filters which are placed on the recessed edge inside the vessel. The caps of the new vessel include an internal O-ring to back up the primary seal formed between the vessel, filter, and the cap. These vessels require changing the side of the vortexer bracket to accommodate the vessel and include a shielded vortexer bearing. These design changes address the known difficulties associated with the assembly of the reaction vessels.
For Research Use Only. Not for use in diagnostics procedures.
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| For Use With (Equipment) | 433 A Peptide Synthesizer |
| For Use With (Application) | Protein Sequencing & Peptide Synthesis |
| Product Type | Peptide Synthesis Vessel |
| Quantity | 40 mL |
| Shipping Condition | Room Temperature |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.