Learn More
Thermo Scientific™ Re-Sealing Tape with 3 layer no adhesive at cavity area
Product Code. 15888611 Shop All Thermo Scientific Products
Change view
Click to view available options
Material:
PET, Silicone, PET
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
15888611 PET, Silicone, PET Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15888611 Supplier Thermo Scientific™ Supplier No. 60180M176
Only null left
Add to basket
Add to basket
Sealing tapes minimize evaporation and protectsamples from contamination and spilling.
Store at room temperature
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material PET, Silicone, PET
Sterility Non-sterile
No. per Pack 25
Product Line Webseal