Learn More
RDM1 Rabbit anti-Human, Polyclonal, Proteintech
Product Code. 16839664 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16839664 20 μL 20µL
16829664 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16839664 Supplier Proteintech Supplier No. 201561AP20UL
Rabbit Polyclonal Antibody
This gene encodes a protein involved in the cellular response to cisplatin, a drug commonly used in chemotherapy. The protein encoded by this gene contains two motifs: a motif found in RAD52, a protein that functions in DNA double-strand breaks and homologous recombination, and an RNA recognition motif (RRM) that is not found in RAD52. The RAD52 motif region in RAD52 is important for protein function and may be involved in DNA binding or oligomerization. Alternatively spliced transcript variants encoding different isoforms have been reported.
Specifications
Antigen RDM1
Applications Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RDM1
Gene Accession No. Q8NG50
Gene Alias RAD52 homolog B, RAD52 motif 1, RAD52B, RDM1
Gene Symbols RDM1
Host Species Rabbit
Immunogen RDM1 Fusion Protein Ag14077
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201299
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG