missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RDH16 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15736301 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15736301 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15736301 Supplier Invitrogen™ Supplier No. PA558423

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GVKVAMIEPG YFKTAVTSKE RFLKSFLEIW DRSSPEVKEA YGEKFVADYK KSAEQMEQKC TQDLSLVT Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 54%.

Retinol dehydrogenase 16 (RDH16) is an oxidoreductase with a preference for NAD. It oxidizes all-trans-retinol and 13-cis-retinol to the corresponding aldehydes. RDH16 has higher activity towards CRBP-bound retinol than with free retinol. It oxidizes 3-alpha-hydroxysteroids and also oxidizes androstanediol and androsterone to dihydrotestosterone and androstanedione. RDH16 can also catalyze the reverse reaction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RDH16
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RDH16
Gene Accession No. O75452
Gene Alias 29 kDa protein; Cis-retinol androgen dehydrogenase 1; cis-retinol/androgen dehydrogenase 1; Crad; CRAD1; hRDH-E; Human epidermal retinol dehydrogenase; Microsomal NAD(+)-dependent retinol dehydrogenase 4; microsomal NAD+-dependent retinol dehydrogenase 4; Rdh1; Rdh16; Rdh2; Rdh6; Retinol dehydrogenase 16; retinol dehydrogenase 16 (all-trans and 13-cis); retinol dehydrogenase 16 (all-trans); retinol dehydrogenase 2; retinol dehydrogenase 6; Retinol dehydrogenase type 1; retinol dehydrogenase type 2; retinol dehydrogenase type 6; Retinol dehydrogenase type II; retinol dehydrogenase type II (RODH II); RODH II; Rodh1; RODH4; RoDH-4; Rodhii; SDR9C8; Short chain dehydrogenase/reductase family 9C member 8; short chain dehydrogenase/reductase family 9C, member 8; sterol/retinol dehydrogenase
Gene Symbols RDH16
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RDH16. Recombinant protein control fragment (Product #RP-97767).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8608
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.