missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
EPLIN is a cytoskeleton-associated protein that inhibits actin filament depolymerization and cross-links filaments in bundles.
Specifications
Specifications
| Antigen | RCOR3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.31 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | RCOR3 |
| Gene Accession No. | Q6PGA0, Q9P2K3 |
| Gene Alias | KIAA1343, RCOR3, REST corepressor 3, RP11 318L16.1 |
| Gene Symbols | RCOR3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?