missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RCL1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16698324
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16698324 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16698324 Supplier Invitrogen™ Supplier No. PA5101854

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RCL1.

RCL1, which does not have cyclase activity, plays a role in 40S- ribosomal-subunit biogenesis in the early pre-rRNA processing steps at sites A0, A1 and A2 that are required for proper maturation of the 18S RNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RCL1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RCL1
Gene Accession No. Q9JJT0, Q9Y2P8
Gene Alias 2310040A02Rik; Ab1-353; Ac1292; AI789745; C76567; HSPC338; Rcl1; RNA 3'-terminal phosphate cyclase-like protein; RNA cyclase homolog; RNA terminal phosphate cyclase like 1; RNA terminal phosphate cyclase-like 1; Rnac; RPC2; RPCL1; RTC2
Gene Symbols RCL1
Host Species Rabbit
Immunogen A synthesized peptide derived from human RCL1(Accession Q9Y2P8), corresponding to amino acid residues I325-K373.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10171, 309301, 59028
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.