missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35137-100ul
This item is not returnable.
View return policy
Description
RBM6 Polyclonal antibody specifically detects RBM6 in Human samples. It is validated for ELISA,Western Blot
Specifications
| RBM6 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| DEF-3, DEF3FLJ39446,3G2, DKFZp686B0877, FLJ36517, FLJ42036, g16, HLC-11, Lung cancer antigen NY-LU-12, lung cancer protooncogene 11, NY-LU-12, Protein G16, RNA binding motif protein 6, RNA-binding motif protein 6, RNA-binding protein 6, RNA-binding protein DEF-3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 530-770 of human RBM6 (NP_005768.1).,, Sequence:, TLEYVSSLDFWYCKRCKANIGGHRSSCSFCKNPREVTEAKQELITYPQPQKTSIPAPLEKQPNQPLRPADKEPEPRKREEGQESRLGHQKREAERYLPPSRREGPTFRRDRERESWSGETRQDGESKTIMLKRIYRSTPPEVIVEVLEPYVRLTTANVRIIKNRTGPMGHTYGFIDLDSHAEALRVVKILQNLDPPFSIDGKMVAVNLATGKRRNDSGDHSDHMHYYQGKKYFRDRRGGGR | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10180 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction