missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-6 Rat anti-Mouse, Clone: MP5-20F3, R&D Systems™

Rat Monoclonal Antibody has been used in 56 publications
Brand: R&D Systems MAB406-100
This item is not returnable.
View return policy
Description
IL-6 Monoclonal specifically detects IL-6 in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| IL-6 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL6 | |
| COS-7 African green monkey SV40 transformed kidney fibroblast-like cell line-derived recombinant mouse IL-6 | |
| 100 μg | |
| Primary | |
| Detects mouse IL-6 in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) IL-6, recombinant porcine IL-6, recombinant rat IL-6, rhIL-11, rhCT-1, or rhCLC is observed. | |
| Mouse | |
| Supernatant |
| Western Blot, Neutralization, ELISA | |
| MP5-20F3 | |
| Western Blot 1 ug/mL, Neutralization 0.005-0.025 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3569 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction