missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASAL2 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-19150AF350
This item is not returnable.
View return policy
Description
RASAL2 Polyclonal antibody specifically detects RASAL2 in Human samples. It is validated for Western Blot
Specifications
| RASAL2 | |
| Polyclonal | |
| Western Blot | |
| MGC129919, NGAP, ras GTPase-activating protein nGAP, Ras protein activator like 1, RAS protein activator like 2, RAS protein activator-like 2 | |
| The immunogen recognized by this antibody maps to a region between residue 888 and 938 of human RAS protein activator like 2 using the numbering given in entry NP_004832.1 (GeneID 9462). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 9462 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction