missing translation for 'onlineSavingsMsg'
Learn More

Scharlab Rappaport Vassiliadis Broth

Product Code. 11411857
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11411857 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11411857 Supplier Scharlab Supplier No. 02379500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rappaport Vassiliadis Broth is used for the selective enrichment of Salmonella in foodstuffs and other samples.

  • AOAC/BAM/COMPF/IDF/ISO
  • Liquid medium for the selective enrichment of Salmonella in foodstuffs and other samples
  • According to ISO 6579, 6785 and FIL-IDF standards.

Specifications

Certifications/Compliance ISO 6579, 6785 and FIL-IDF
Form Liquid
Media Type Rappaport Vassiliadis Broth
Product Type Microbiology Media
Target Organism Class Gram-negative Bacteria
Formulation Soy Peptone 4.50 g/L, Sodium Chloride 7.20 g/L, Monopotassium Phosphate 1.26 g/L, Dipotassium Phosphate 0.18 g/L, Magnesium Chloride (Anhydrous) 13.40 g/L, Malachite Green 0.036 g/L
For Use With (Application) Selective Enrichment of Salmonella in Foodstuffs
Incubation Conditions 41.5°C ± 1°C
Incubation Time 24 ± 3 hr.
pH 5.2 ±0.2
Quantity 500 g
Species Salmonella species
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.