missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rapastinel Trifluoroacetate, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
Description
Rapastinel Trifluoroacetate (GLYX-13 Trifluoroacetate) is an NMDA receptor modulator with glycine-site partial agonist properties. Rapastinel Trifluoroacetate has the potential for major depressive disorder treatment.
Specifications
Specifications
| Chemical Name or Material | Rapastinel Trifluoroacetate |
| CAS | 1435786-04-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C20H32F3N5O8 |
| Quantity | 1 mg |
| Synonym | GLYX-13 Trifluoroacetate |
| Solubility Information | DMSO : 125 mg/mL (236.97 mM; Need ultrasonic) |
| SMILES | O=C([C@H]1N(C([C@H]2N(C([C@@H](N)[C@H](O)C)=O)CCC2)=O)CCC1)N[C@@H]([C@H](O)C)C(N)=O.O=C(O)C(F)(F)F |
| Molecular Weight (g/mol) | 527.49 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction