Learn More
R&D Systems™ Stop Solution 2N Sulfuric Acid
Product Code. 18754424
Click to view available options
Quantity:
15 x 6 mL
Unit Size:
90mL
Product Code. 18754424
Brand: R&D Systems™ DY994
For stopping the action of HRP on the substrate, tetramethylbenzidine
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) ELISA
Quantity 15 x 6 mL