Learn More
Mettler Toledo™ Rainin™ Bioclean™ Pipette Tip, Packaged in Racks
Click to view available options
Volume (Metric):
20 μL
300 μL
Unit Size:
Pack of 768
Pack of 960
Specifications
Specifications
| Volume (Metric) | 20 μL |
| For Use With (Application) | Perfect production and advanced design |
| Format | Rack |
| For Use With (Equipment) | Rainin |
| Product Line | Rainin |