missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ Rainin™ Bioclean™ Pipette Tip, Packaged in Racks
Specifications
Specifications
| Volume (Metric) | 300 μL |
| For Use With (Application) | Perfect production and advanced design |
| Format | Rack |
| For Use With (Equipment) | Rainin |
| Product Line | Rainin BioClean |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?