Learn More
Radiprodil, MedChemExpress
Product Code. 16466546
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16466546
Brand: Medchem Express HY14777
MedChemExpress Radiprodil (RGH-896) is an orally active and selective NMDA NR2B antagonist. A potential therapeutic agent in treatment of neuropathic pain and possibly other chronic pain conditions.
Radiprodil (RGH-896) is an orally active and selective NMDA NR2B antagonist. A potential therapeutic agent in treatment of neuropathic pain and possibly other chronic pain conditions.
Specifications
Chemical Name or Material Radiprodil
CAS 496054-87-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H20FN3O4
Quantity 100 mg
Synonym RGH-896
Solubility Information DMSO : 250 mg/mL (629.09 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C2NC(OC2=C1)=O)C(N3CCC(CC4=CC=C(F)C=C4)CC3)=O
Molecular Weight (g/mol) 397.4
Formula Weight 397.4
Percent Purity 99.06%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration