missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAD51 Recombinant Rabbit Monoclonal Antibody (5S9B0)
GREENER_CHOICE

Product Code. 17136023
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17136023 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17136023 Supplier Invitrogen™ Supplier No. MA535363

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MAMQMQLEAN ADTSVEEESF GPQPISRLEQ CGINANDVKK LEEAGFHTVE AVAYAPKKEL INIKGISEAK ADKILAEAAK LVPMGFTTAT EFHQRRSEII.

RAD51 plays a critical role in homologous strand exchange, a key step in DNA repair through homologous recobination. It binds to single and double-stranded DNA and exhibits DNA-dependent ATPase activity. RAD51 catalyzes the recognition of homology and strand exchange between homologous DNA partners to form a joint molecule between a processed DNA break and the repair template. It binds to a single-stranded DNA in an ATP-dependent manner to form nucleoprotein filaments which are essential for the homology search and strand exchange. Mutations in the gene can results in breast cancer, mirror movements 2 and fanconi anemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAD51
Applications ChIP Assay, ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 5S9B0
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3
Gene Rad51
Gene Accession No. Q06609, Q08297
Gene Alias AV304093; BRCA1/BRCA2-containing complex, subunit 5; BRCC5; DNA repair protein RAD51; DNA repair protein RAD51 homolog 1; DNA repair protein RAD51 homolog A; DNA repair protein rhp51; FANCR; homolog to S.cerevisiae; HRAD51; HsRad51; HsT16930; MGC128559 protein; MRMV2; MUT5; rad51; RAD51 homolog; RAD51 homolog (RecA homolog); RAD51 homolog (RecA homolog, E. coli); RAD51 homolog A; RAD51 recombinase; RAD51 recombinase L homeolog; rad51.1; rad51.L; rad51-1; RAD51A; rad51-a; rad51-b; Reca; RecA family recombinase Rhp51; RecA, E. coli, homolog of; RecA-like protein; recombinase RAD51; recombination protein A; RGD1563603; rhp51; SPAC644.14c; wu:fb38e12; xrad51; XRad51.1; YER095W; zgc:77754
Gene Symbols Rad51
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19361, 499870, 5888
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.