Learn More
Thermo Scientific™ Specialty Racks
Product Code. p-4520827 Shop All Thermo Scientific Products
Click to view available options
Holds:
Boxes (2 in.)
Boxes (3 in.)
General Freezer Items
Microplates
Microplates or Deepwell Microplates
Positions:
12 Boxes/Rack
15 Boxes/Rack
20 Boxes/Rack
22 Boxes/Rack
36 Plates/Rack
56 Plates/Rack
77 Plates/Rack
84 Plates/Rack
9 Boxes/Rack
98 microplates with lids, 119 microplates without lids, and 35 deepwell
Product Type:
Box Racks - Sliding Drawer
Side Access Freezer Rack (Adjustable)
Side Access Freezer Rack (Microplate/Deepwell)
Side Access Freezer Rack (Microplate/Locking Rod)
Specialty Rack - 8-Compartment Container
For Use With (Equipment):
13 cu. ft. Upright Freezers
17.3, 23 cu. ft. Upright Freezers
32 cu.ft. Revco Upright ULT
Thermo Scientific TDE and TSX Series (5 shelves)
Unit Size:
Each
Product Code. 12662725
Brand: Thermo Scientific™ 1950651
Only null left
Add to basket
Add to basket
Maximize the storage space in your ultra-low temperature freezer with these racks.
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for previous generation Forma™ 900, 7000 Series, Revco™ UxF, HERAfreeze™ HFU T and TSE/TSD Series Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
  • With sliding drawer racks
  • Adjustable side access box racks
  • Microplate racks in standard or deepwell formats
TRUSTED_SUSTAINABILITY
Specifications
Product Type Side Access Freezer Rack (Microplate/Deepwell)
Color Silver
Holds Microplates
Material Stainless Steel
Positions 56 Plates/Rack
Shape Square
For Use With (Equipment) 13 cu. ft. Upright Freezers
Width (Metric) 14.0 cm
Width (English) 5.5 in.
Height (Metric) 30.2 cm
Height (English) 11.9 in.
Depth (English) 14.7 in.
Depth (Metric) 37.3 cm