Learn More
Thermo Scientific™ Specialty Racks
Shop All Thermo Scientific Products
Click to view available options
Holds:
Boxes (2 in.)
Boxes (3 in.)
General Freezer Items
Microplates
Microplates or Deepwell Microplates
Positions:
12 Boxes/Rack
15 Boxes/Rack
20 Boxes/Rack
22 Boxes/Rack
36 Plates/Rack
56 Plates/Rack
77 Plates/Rack
84 Plates/Rack
9 Boxes/Rack
98 microplates with lids, 119 microplates without lids, and 35 deepwell
Product Type:
Box Racks - Sliding Drawer
Side Access Freezer Rack (Adjustable)
Side Access Freezer Rack (Microplate/Deepwell)
Side Access Freezer Rack (Microplate/Locking Rod)
Specialty Rack - 8-Compartment Container
For Use With (Equipment):
13 cu. ft. Upright Freezers
17.3, 23 cu. ft. Upright Freezers
32 cu.ft. Revco Upright ULT
Thermo Scientific TDE and TSX Series (5 shelves)
Unit Size:
Each
Description
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for previous generation Forma™ 900, 7000 Series, Revco™ UxF, HERAfreeze™ HFU T and TSE/TSD Series Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
- Warranty varies by country, please contact us for details
Specifications
Specifications
| Product Type | Side Access Freezer Rack (Microplate/Locking Rod) |
| Color | Silver |
| Holds | Microplates or Deepwell Microplates |
| Material | Stainless Steel |
| Positions | 98 microplates with lids, 119 microplates without lids, and 35 deepwell |
| Shape | Rectangular |
| Shelves | 4 Tiers |
| For Use With (Equipment) | 32 cu.ft. Revco Upright ULT |
| Width (Metric) | 14 cm |
| Width (English) | 5.5 in. |
| Show More |