Learn More
asecos™ Rack sump PE for Mounting with PE Grid
Product Code. p-200003662 Shop All asecos Products
Click to view available options
Width (Metric):
1780.00 mm
2680.00 mm
Unit Size:
Each
Product Code. 17886566
Brand: asecos™ 22459
This item requires a price update prior to being ordered.
Request a Price for this item here.
The PE Rack Sump is approved for the safe storage of water pollutants.
  • Rack sump
  • PE blue
  • Grid: PE
  • >
  • Ideal for wall mounting
  • Sump capacity: 225 litres
  • Load capacity in kg: 800
Specifications
Color Blue
Capacity (English) 59.4 gal.
Capacity (Metric) 225 L
Material Polyethylene
Type Sump
Length (English) 70.08 in.
Length (Metric) 1780.00 mm
Width (English) 70.08 in.
Width (Metric) 1780.00 mm
Height (English) 10.83 in.
Height (Metric) 275.00 mm
Depth (English) 51.18 in.
Depth (Metric) 1300.00 mm
Weight (English) 92.59 lb.
Weight (Metric) 42 kg