Learn More
Tenak™ Comfort Rack for Microtiter Plates for Upright Freezers, 25mm Shelf Height
Shop All TENAK Products
Click to view available options
Holds:
Plates (24)
Plates (30)
Plates (32)
Plates (36)
Plates (40)
Plates (42)
Plates (48)
Plates (50)
Plates (60)
Plates (70)
Plates (72)
Plates (75)
Plates (84)
Dimensions (L x W x H):
360 x 135 x 159 mm
360 x 135 x 212 mm
360 x 135 x 265 mm
360 x 135 x 318 mm
450 x 135 x 159 mm
450 x 135 x 212 mm
450 x 135 x 265 mm
450 x 135 x 318 mm
450 x 135 x 371 mm
450 x 135 x 398 mm
540 x 135 x 159 mm
540 x 135 x 212 mm
540 x 135 x 265 mm
540 x 135 x 318 mm
630 x 135 x 159 mm
630 x 135 x 318 mm
Unit Size:
Each
Description
- Racks can either be used as one plate per shelf or the plates can be stacked
- With an alert preventing the shelves from sliding out
- External footprint is 135 x 92mm
Specifications
Specifications
| Product Type | Microtiter Plate Rack |
| Array | 8 x 5 |
| Holds | Plates (40) |
| Shelves | 40 |
| Stackable | Yes |
| For Use With (Equipment) | Microtiter Plates, Upright Freezers |
| No. per Pack | 1 Pack |
| Dimensions (L x W x H) | 450 x 135 x 212 mm |
| Width (Metric) | 135 mm |
| Height (Metric) | 212 mm |
| Show More |